Assay Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199253.2 | NP_954862.2 | 4 - 5 | 847 | 144 | |
| GenBank mRNA | AY847775.1 | - | 4 - 5 | 847 | 144 | |
| BC133063.1 | - | 4 - 5 | 778 | 8371686 | 144 | |
More Information
Set Membership: |
|

Pcsk9
Genomic Map