Assay Details
| Gene Symbol: Btbd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729514.4 | XP_002729560.1 | 4 - 5 | 732 | 83 | |
| XM_006238729.2 | XP_006238791.1 | 4 - 5 | 732 | 83 | ||
| XM_006238730.2 | XP_006238792.1 | 3 - 4 | 850 | 83 | ||
| XM_008764072.1 | XP_008762294.1 | 3 - 4 | 852 | 83 | ||
| XM_008764075.1 | XP_008762297.1 | 1 - 2 | 1117 | 83 | ||
More Information
Set Membership: |
|

Btbd19
Genomic Map