Assay Details
| Gene Symbol: Btbd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729514.4 | XP_002729560.1 | 5 - 6 | 808 | 69 | |
| XM_006238729.2 | XP_006238791.1 | 5 - 6 | 808 | 69 | ||
| XM_006238730.2 | XP_006238792.1 | 4 - 5 | 926 | 69 | ||
| XM_008764072.1 | XP_008762294.1 | 4 - 5 | 928 | 69 | ||
| XM_008764073.1 | XP_008762295.1 | 3 - 4 | 868 | 69 | ||
| XM_008764074.1 | XP_008762296.1 | 4 - 5 | 748 | 69 | ||
| XM_008764075.1 | XP_008762297.1 | 2 - 3 | 1193 | 69 | ||
More Information
Set Membership: |
|

Btbd19
Genomic Map