Assay Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100840.1 | NP_001094310.1 | 4 - 5 | 425 | 112 | |
| GenBank mRNA | BC100255.1 | - | 4 - 5 | 367 | 112 | |
| BC158737.1 | - | 4 - 5 | 371 | 112 | ||
| CV117277.1 | - | 4 - 5 | 662 | 112 | ||
| FQ218354.1 | - | 4 - 5 | 322 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txndc12
Genomic Map