Assay Details
| Gene Symbol: Mroh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100965.2 | NP_001094435.1 | 14 - 15 | 2684 | 62 | |
| XM_008763860.1 | XP_008762082.1 | 16 - 17 | 2943 | 62 | ||
| XM_008763861.1 | XP_008762083.1 | 15 - 16 | 2872 | 62 | ||
| XM_008763862.1 | XP_008762084.1 | 16 - 17 | 2844 | 62 | ||
| XM_008763863.1 | XP_008762085.1 | 16 - 17 | 3226 | 62 | ||
| XM_008763864.1 | XP_008762086.1 | 15 - 16 | 2773 | 62 | ||
| XM_008763865.1 | XP_008762087.1 | 15 - 16 | 2701 | 62 | ||
| XM_008763866.1 | XP_008762088.1 | 16 - 17 | 3015 | 62 | ||
| GenBank mRNA | BC133066.1 | - | 14 - 15 | 2687 | 7134332 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Mroh7
Genomic Map