Assay Details
| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107963.1 | NP_001101433.1 | 11 - 12 | 2002 | 79 | |
| XM_008764009.1 | XP_008762231.1 | 11 - 12 | 1924 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim5
Genomic Map