Assay Details
| Gene Symbol: Cyp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044770.2 | NP_001038235.1 | 4 - 5 | 508 | 69 | |
| GenBank mRNA | BC078684.1 | - | 4 - 5 | 508 | 7109305 | 69 |
| BC098705.1 | - | 4 - 5 | 503 | 7106510 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Cyp4a2
Genomic Map