Assay Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729532.4 | XP_002729578.2 | 3 - 4 | 465 | 86 | |
| XM_006238746.2 | XP_006238808.1 | 3 - 4 | 467 | 86 | ||
| XM_006238747.2 | XP_006238809.1 | 3 - 4 | 467 | 86 | ||
| XM_006238748.2 | XP_006238810.1 | 3 - 4 | 467 | 86 | ||
More Information
Set Membership: |
|

Hyi
Genomic Map