Assay Details
| Gene Symbol: Btf3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_345561.7 | XP_345562.4 | 1 - 2 | 112 | 95 | |
| GenBank mRNA | FQ222777.1 | - | 1 - 2 | 109 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_345561.7 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Btf3l4
Genomic Map