Assay Details
| Gene Symbol: Lingo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766270.1 | XP_008764492.1 | 1 - 2 | 386 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766270.1 transcripts.
Set Membership: |
|

Lingo1
Genomic Map