Assay Details
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053297.2 | NP_445749.1 | 1 - 2 | 68 | 69 | |
| XM_006243187.2 | XP_006243249.1 | 1 - 2 | 298 | 69 | ||
| XM_006243188.2 | XP_006243250.1 | 1 - 2 | 298 | 69 | ||
| GenBank mRNA | AY724474.1 | - | 1 - 2 | 65 | 69 | |
| BC061541.1 | - | 1 - 2 | 75 | 6888187 | 69 | |
| FQ223721.1 | - | 1 - 2 | 76 | 69 | ||
| X15800.1 | - | 1 - 2 | 70 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061541.1, NM_053297.2, X15800.1 transcripts.
Set Membership: |
|

Pkm
Genomic Map