Assay Details
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053297.2 | NP_445749.1 | 4 - 5 | 460 | 98 | |
| XM_006243187.2 | XP_006243249.1 | 4 - 5 | 690 | 98 | ||
| XM_006243188.2 | XP_006243250.1 | 4 - 5 | 690 | 98 | ||
| XM_006243190.2 | XP_006243252.1 | 5 - 6 | 762 | 98 | ||
| GenBank mRNA | AY724474.1 | - | 4 - 5 | 457 | 98 | |
| BC061541.1 | - | 4 - 5 | 467 | 6888187 | 98 | |
| FQ223721.1 | - | 4 - 5 | 468 | 98 | ||
| FQ224316.1 | - | 1 - 2 | 118 | 98 | ||
| X15800.1 | - | 4 - 5 | 462 | 98 | ||
| Gene Symbol: LOC100364062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 384 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pkm
Genomic Map