Assay Details
| Gene Symbol: LOC100364062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 1145 | 84 | |
| Gene Symbol: LOC100362738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 1185 | 84 | |
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243188.2 | XP_006243250.1 | 8 - 9 | 1451 | 84 | |
| GenBank mRNA | AY724474.1 | - | 8 - 9 | 1218 | 84 | |
| BC061541.1 | - | 8 - 9 | 1096 | 6888187 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100364062
Genomic Map