Assay Details
| Gene Symbol: Sema7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108153.1 | NP_001101623.1 | 10 - 11 | 1298 | 114 | |
| XM_008766273.1 | XP_008764495.1 | 10 - 11 | 1145 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema7a
Genomic Map