Assay Details
| Gene Symbol: Ppcdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108763.1 | NP_001102233.1 | 3 - 4 | 346 | 75 | |
| XM_008766303.1 | XP_008764525.1 | 3 - 4 | 438 | 75 | ||
| XM_008766304.1 | XP_008764526.1 | 4 - 5 | 1919 | 75 | ||
| XM_008766305.1 | XP_008764527.1 | 4 - 5 | 473 | 75 | ||
| XM_008766306.1 | XP_008764528.1 | 4 - 5 | 497 | 75 | ||
| XM_008766307.1 | XP_008764529.1 | 5 - 6 | 1199 | 75 | ||
| Gene Symbol: NEWGENE_1306267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750512.1 | XP_003750560.1 | 344 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppcdc
Genomic Map