Assay Details
| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008880.1 | NP_001008880.1 | 3 - 4 | 462 | 63 | |
| GenBank mRNA | AF544988.1 | - | 1 - 2 | 65 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scn4b
Genomic Map