Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080756.1 | NP_001074225.1 | 4 - 5 | 434 | 82 | |
| XM_006243310.2 | XP_006243372.1 | 4 - 5 | 588 | 82 | ||
| XM_006243311.2 | XP_006243373.1 | 4 - 5 | 588 | 82 | ||
| GenBank mRNA | AY952140.1 | - | 4 - 5 | 398 | 82 | |
| EF187255.1 | - | 4 - 5 | 434 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car12
Genomic Map