Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080756.1 | NP_001074225.1 | 9 - 10 | 910 | 123 | |
| XM_006243310.2 | XP_006243372.1 | 9 - 10 | 1067 | 126 | ||
| GenBank mRNA | EF187255.1 | - | 9 - 10 | 910 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car12
Genomic Map