Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243310.2 | XP_006243372.1 | 10 - 11 | 1148 | 138 | |
| XM_006243311.2 | XP_006243373.1 | 9 - 10 | 1115 | 138 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P027373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105785 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027373 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027373 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027373 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P424529 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104132 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100119 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10911154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911151 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911149 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911155 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911148 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911147 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911150 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10911145 | - | - |
| Rat Genome 230 2.0 Array | 1391198_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391198_at | - | - |
| Rat Genome 230 2.0 Array | 1371922_at | - | - |
| Rat Genome 230 2.0 Array | 1371922_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391198_at | - | - |
| Affymetrix HT Rat Focus Array | 1371922_at | - | - |
| Affymetrix HT Rat Focus Array | 1371922_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391198_PM_at | - | - |
| Rat Expression Set 230 | 1371922_PM_at | - | - |
| Rat Expression Set 230 | 1371922_at | - | - |
| Rat Expression Set 230 | 1391198_at | - | - |
| Rat Expression Set 230 | 1391198_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911145 | - | - |
| Rat Genome U34 Set | rc_AI233771_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21425311 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354371 | - | - |

Car12
Genomic Map