Assay Details
| Gene Symbol: Lrrc49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134469.1 | NP_001127941.1 | 7 - 8 | 749 | 61 | |
| XM_006243202.2 | XP_006243264.1 | 8 - 9 | 1221 | 61 | ||
| XM_006243204.2 | XP_006243266.1 | 7 - 8 | 1165 | 61 | ||
| GenBank mRNA | BC160848.1 | - | 7 - 8 | 749 | 8363379 | 61 |
| FQ211943.1 | - | 6 - 7 | 902 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc49
Genomic Map