Assay Details
| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017078.2 | NP_058774.2 | 2 - 3 | 311 | 92 | |
| GenBank mRNA | AY574255.1 | - | 2 - 3 | 209 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrna5
Genomic Map