Assay Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016989.2 | NP_058685.1 | 1 - 2 | 294 | 62 | |
| XM_006245672.2 | XP_006245734.1 | 2 - 3 | 990 | 62 | ||
| XM_006245673.2 | XP_006245735.1 | 2 - 3 | 1366 | 62 | ||
| XM_006245674.2 | XP_006245736.1 | 2 - 3 | 1319 | 62 | ||
| XM_008767418.1 | XP_008765640.1 | 2 - 3 | 943 | 62 | ||
| GenBank mRNA | AF252410.1 | - | 2 - 3 | 194 | 62 | |
| CB786482.1 | - | 1 - 2 | 306 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF252410.1, NM_016989.2, XM_006245672.2, XM_006245673.2, XM_006245674.2, XM_008767418.1 transcripts.
Set Membership: |
|

Adcyap1
Genomic Map