Assay Details
| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013017.1 | NP_037149.1 | 578 | 75 | ||
| GenBank mRNA | DY310463.1 | - | 99 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab12
Genomic Map