Assay Details
| Gene Symbol: Efnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017089.2 | NP_058785.2 | 2 - 3 | 1156 | 113 | |
| GenBank mRNA | BC061744.1 | - | 2 - 3 | 1156 | 5599058 | 113 |
| U07560.1 | - | 2 - 3 | 1182 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efnb1
Genomic Map