Assay Details
| Gene Symbol: Tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752051.3 | XP_003752099.1 | 1 - 2 | 2128 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003752051.3 transcripts.
Set Membership: |
|

Tab3
Genomic Map