Assay Details
| Gene Symbol: Gjb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017251.2 | NP_058947.1 | 1 - 2 | 74 | 62 | |
| GenBank mRNA | BC078868.1 | - | 1 - 2 | 38 | 7127952 | 62 |
| CK476084.1 | - | 1 - 2 | 74 | 62 | ||
| FQ210131.1 | - | 1 - 2 | 59 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078868.1, NM_017251.2 transcripts.
Set Membership: |
|

Gjb1
Genomic Map