Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053512.2 | NP_445964.1 | 4 - 5 | 629 | 104 | |
| XM_006256951.2 | XP_006257013.1 | 4 - 5 | 794 | 104 | ||
| GenBank mRNA | AF106945.1 | - | 4 - 5 | 630 | 104 | |
| BC059122.1 | - | 4 - 5 | 713 | 6918063 | 104 | |
| FQ219258.1 | - | 4 - 5 | 677 | 104 | ||
| FQ220216.1 | - | 4 - 5 | 683 | 104 | ||
| FQ220257.1 | - | 4 - 5 | 677 | 104 | ||
| FQ220425.1 | - | 4 - 5 | 678 | 104 | ||
| FQ220916.1 | - | 4 - 5 | 678 | 104 | ||
| FQ223235.1 | - | 4 - 5 | 673 | 104 | ||
| FQ228535.1 | - | 4 - 5 | 678 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdx4
Genomic Map