Assay Details
| Gene Symbol: Kif4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257113.2 | XP_006257175.1 | 2 - 3 | 389 | 143 | |
| XM_006257114.1 | XP_006257176.1 | 2 - 3 | 405 | 143 | ||
| XM_343797.7 | XP_343798.2 | 1 - 2 | 504 | 143 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif4a
Genomic Map