Assay Details
| Gene Symbol: Kif4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257113.2 | XP_006257175.1 | 26 - 27 | 3249 | 118 | |
| XM_006257114.1 | XP_006257176.1 | 26 - 27 | 3265 | 118 | ||
| XM_343797.7 | XP_343798.2 | 25 - 26 | 3364 | 118 | ||
More Information
Set Membership: |
|

Kif4a
Genomic Map