Assay Details
| Gene Symbol: Eif2s3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100542.1 | NP_001094012.1 | 9 - 10 | 1035 | 152 | |
| GenBank mRNA | BC091286.1 | - | 7 - 8 | 818 | 152 | |
| BC166783.1 | - | 9 - 10 | 1017 | 7935021 | 152 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif2s3x
Genomic Map