Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108874.2 | NP_001102344.1 | 7 - 8 | 665 | 64 | |
| XM_006252913.1 | XP_006252975.1 | 6 - 7 | 717 | 64 | ||
| XM_006252914.1 | XP_006252976.1 | 5 - 6 | 420 | 64 | ||
| XM_008771143.1 | XP_008769365.1 | 6 - 7 | 685 | 64 | ||
| GenBank mRNA | BC161989.1 | - | 7 - 8 | 665 | 9032206 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem161a
Genomic Map