Assay Details
| Gene Symbol: Crlf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106074.1 | NP_001099544.1 | 4 - 5 | 827 | 128 | |
| XM_006252872.2 | XP_006252934.1 | 4 - 5 | 870 | 128 | ||
| XM_006252874.2 | XP_006252936.1 | 4 - 5 | 870 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crlf1
Genomic Map