Assay Details
| Gene Symbol: Cyp4f18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033686.1 | NP_001028858.1 | 369 | 82 | ||
| GenBank mRNA | BC101918.1 | - | 369 | 7935301 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp4f18
Genomic Map