Assay Details
| Gene Symbol: Cyp4f18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033686.1 | NP_001028858.1 | 1275 | 90 | ||
| GenBank mRNA | BC101918.1 | - | 1275 | 7935301 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
leukotriene B4 catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
heme binding
leukotriene-B4 20-monooxygenase activity
alpha-tocopherol omega-hydroxylase activity
leukotriene B4 catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
heme binding
leukotriene-B4 20-monooxygenase activity
alpha-tocopherol omega-hydroxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P051518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168917 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051518 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051518 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P051518 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103528 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99285 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79989 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103841 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10787208 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787205 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10787201 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10787197 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600002303.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600002299.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600002297.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600002301.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600002296.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600002302.rn.1 | TC1600000237.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10787197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21246932 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361303 | - | - |

Cyp4f18
Genomic Map