Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134796.1 | NP_001128268.1 | 19 - 20 | 2390 | 88 | |
| XM_006252943.2 | XP_006253005.1 | 20 - 21 | 2851 | 88 | ||
| XM_006252944.2 | XP_006253006.1 | 20 - 21 | 6382 | 88 | ||
| XM_006252945.2 | XP_006253007.1 | 21 - 22 | 2289 | 88 | ||
| XM_006252946.2 | XP_006253008.1 | 19 - 20 | 6358 | 88 | ||
| XM_006252947.2 | XP_006253009.1 | 20 - 21 | 2268 | 88 | ||
| XM_008771150.1 | XP_008769372.1 | 21 - 22 | 2872 | 88 | ||
| XM_008771151.1 | XP_008769373.1 | 21 - 22 | 2869 | 88 | ||
| XM_008771152.1 | XP_008769374.1 | 21 - 22 | 2857 | 88 | ||
| XM_008771153.1 | XP_008769375.1 | 20 - 21 | 2848 | 88 | ||
| XM_008771154.1 | XP_008769376.1 | 19 - 20 | 6358 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mast3
Genomic Map