Assay Details
| Gene Symbol: Sftpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012878.2 | NP_037010.1 | 6 - 7 | 768 | 85 | |
| XM_008771065.1 | XP_008769287.1 | 6 - 7 | 771 | 85 | ||
| GenBank mRNA | BC070507.1 | - | 7 - 8 | 777 | 7098194 | 85 |
| M81231.1 | - | 7 - 8 | 775 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sftpd
Genomic Map