Assay Details
| Gene Symbol: Cib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191081.1 | NP_001178010.1 | 4 - 5 | 350 | 69 | |
| XM_008771148.1 | XP_008769370.1 | 2 - 3 | 203 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cib3
Genomic Map