Assay Details
| Gene Symbol: Map1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106070.1 | NP_001099540.1 | 4 - 5 | 479 | 61 | |
| XM_008771085.1 | XP_008769307.1 | 4 - 5 | 818 | 61 | ||
| XM_008771086.1 | XP_008769308.1 | 5 - 6 | 564 | 61 | ||
| GenBank mRNA | BC169058.1 | - | 4 - 5 | 451 | 7932017 | 61 |
| FQ211658.1 | - | 4 - 5 | 488 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map1s
Genomic Map