Assay Details
| Gene Symbol: LOC100911483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277222.1 | NP_001264151.1 | 1 - 2 | 92 | 70 | |
| GenBank mRNA | FM065403.1 | - | 1 - 2 | 88 | 70 | |
| FQ211515.1 | - | 1 - 2 | 160 | 70 | ||
| FQ223566.1 | - | 1 - 2 | 103 | 70 | ||
| FQ223934.1 | - | 1 - 2 | 117 | 70 | ||
| FQ224478.1 | - | 1 - 2 | 105 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911483
Genomic Map