Assay Details
| Gene Symbol: Cilp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107307.1 | NP_001100777.1 | 6 - 7 | 1054 | 59 | |
| XM_006252902.2 | XP_006252964.1 | 6 - 7 | 1063 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cilp2
Genomic Map