Assay Details
| Gene Symbol: Mef2bnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145726.1 | NP_001139198.1 | 1 - 2 | 66 | 68 | |
| GenBank mRNA | CB782819.1 | - | 1 - 2 | 86 | 68 | |
| DV714338.1 | - | 1 - 2 | 49 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2bnb
Genomic Map