Assay Details
| Gene Symbol: Mef2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017507.1 | NP_001017507.1 | 2 - 3 | 118 | 71 | |
| XM_006252927.2 | XP_006252989.1 | 2 - 3 | 438 | 71 | ||
| GenBank mRNA | BC079361.1 | - | 2 - 3 | 118 | 7109152 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Mef2b
Genomic Map