Assay Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773209.1 | XP_008771431.1 | 4 - 5 | 765 | 57 | |
| XM_008773210.1 | XP_008771432.1 | 4 - 5 | 762 | 57 | ||
More Information
Set Membership: |
|

Reps2
Genomic Map