Assay Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025416.1 | NP_001020587.1 | 1 - 2 | 65 | 80 | |
| GenBank mRNA | BC097972.1 | - | 1 - 2 | 65 | 7442066 | 80 |
More Information
Updated Mapping Information
Set Membership: |
|

Dok1
Genomic Map