Assay Details
| Gene Symbol: Tacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012667.2 | NP_036799.1 | 1292 | 66 | ||
| GenBank mRNA | J05097.1 | - | 1292 | 66 | ||
| M31477.1 | - | 3 - 4 | 749 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
aggressive behavior
acute inflammatory response
positive regulation of leukocyte migration
angiotensin-mediated drinking behavior
positive regulation of cytosolic calcium ion concentration
tachykinin receptor signaling pathway
neuropeptide signaling pathway
learning or memory
long-term memory
regulation of blood pressure
associative learning
response to heat
response to hormone
response to ozone
positive regulation of epithelial cell migration
response to auditory stimulus
response to organic cyclic compound
regulation of smooth muscle cell migration
sensory perception of pain
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
response to estradiol
response to progesterone
response to nicotine
operant conditioning
positive regulation of renal sodium excretion
sperm ejaculation
eating behavior
positive regulation of vascular permeability
response to morphine
response to ethanol
positive regulation of action potential
positive regulation of blood pressure
positive regulation of ossification
positive regulation of vasoconstriction
positive regulation of saliva secretion
positive regulation of hormone secretion
response to pain
behavioral response to pain
regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
positive regulation of epithelial cell proliferation
positive regulation of stress fiber assembly
response to electrical stimulus
smooth muscle contraction involved in micturition
positive regulation of uterine smooth muscle contraction
substance P receptor activity
aggressive behavior
acute inflammatory response
positive regulation of leukocyte migration
angiotensin-mediated drinking behavior
positive regulation of cytosolic calcium ion concentration
tachykinin receptor signaling pathway
neuropeptide signaling pathway
learning or memory
long-term memory
regulation of blood pressure
associative learning
response to heat
response to hormone
response to ozone
positive regulation of epithelial cell migration
response to auditory stimulus
response to organic cyclic compound
regulation of smooth muscle cell migration
sensory perception of pain
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
response to estradiol
response to progesterone
response to nicotine
operant conditioning
positive regulation of renal sodium excretion
sperm ejaculation
eating behavior
positive regulation of vascular permeability
response to morphine
response to ethanol
positive regulation of action potential
positive regulation of blood pressure
positive regulation of ossification
positive regulation of vasoconstriction
positive regulation of saliva secretion
positive regulation of hormone secretion
response to pain
behavioral response to pain
regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
positive regulation of epithelial cell proliferation
positive regulation of stress fiber assembly
response to electrical stimulus
smooth muscle contraction involved in micturition
positive regulation of uterine smooth muscle contraction
substance P receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P215075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99081 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P215075 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P215075 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P215075 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P215075 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11366 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68934 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67090 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93590 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61134 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36466 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48579 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36221 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23114 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10856530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856531 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856527 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10856525 | - | - |
| Rat Genome 230 2.0 Array | 1387474_at | - | - |
| Rat Genome 230 2.0 Array | 1387474_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369199_at | - | - |
| Rat Genome 230 2.0 Array | 1369199_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369199_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369199_at | - | - |
| Affymetrix HT Rat Focus Array | 1387474_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387474_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400008463.rn.1 | TC0400001215.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400008464.rn.1 | TC0400001215.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387474_PM_at | - | - |
| Rat Expression Set 230 | 1369199_PM_at | - | - |
| Rat Expression Set 230 | 1387474_at | - | - |
| Rat Expression Set 230 | 1369199_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856525 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21063330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376350 | - | - |

Tacr1
Genomic Map