Assay Details
| Gene Symbol: Tada3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025734.1 | NP_001020905.1 | 5 - 6 | 1012 | 67 | |
| XM_006237038.1 | XP_006237100.1 | 6 - 7 | 1479 | 67 | ||
| XM_006237040.1 | XP_006237102.1 | 6 - 7 | 1305 | 67 | ||
| XM_006237042.1 | XP_006237104.1 | 5 - 6 | 1251 | 67 | ||
| XM_006237043.1 | XP_006237105.1 | 6 - 7 | 1479 | 67 | ||
| GenBank mRNA | BC097414.1 | - | 5 - 6 | 1012 | 7454313 | 67 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tada3
Genomic Map