Assay Details
| Gene Symbol: Cd8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031539.1 | NP_113727.1 | 1 - 2 | 80 | 72 | |
| GenBank mRNA | BC059127.1 | - | 1 - 2 | 102 | 6888323 | 72 |
| X04310.1 | - | 1 - 2 | 80 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd8b
Genomic Map