Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134554.1 | NP_001128026.1 | 13 - 14 | 1805 | 79 | |
| XM_006236702.2 | XP_006236764.1 | 14 - 15 | 1986 | 79 | ||
| XM_006236703.2 | XP_006236765.1 | 14 - 15 | 1986 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcfc2
Genomic Map