Assay Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014188.1 | NP_001014210.1 | 3 - 4 | 160 | 71 | |
| GenBank mRNA | BC079119.1 | - | 3 - 4 | 160 | 7126525 | 71 |
| FQ218201.1 | - | 3 - 4 | 211 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isy1
Genomic Map