Assay Details
| Gene Symbol: Rybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107879.1 | NP_001101349.1 | 3 - 4 | 401 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107879.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P126955 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P758991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118213 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126955 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P758991 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126955 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P758991 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P758991 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P126955 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P620448 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P758991 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10052 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31697 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10864444 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864442 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864443 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864445 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10864441 | - | - |
| Rat Genome 230 2.0 Array | 1383034_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372674_at | - | - |
| Rat Genome 230 2.0 Array | 1372674_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383034_at | - | - |
| Affymetrix HT Rat Focus Array | 1372674_at | - | - |
| Affymetrix HT Rat Focus Array | 1372674_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383034_at | - | - |
| Affymetrix HT Rat Focus Array | 1383034_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400024400.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024404.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024398.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024399.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024403.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024401.rn.1 | TC0400003459.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400024402.rn.1 | TC0400003459.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372674_at | - | - |
| Rat Expression Set 230 | 1372674_PM_at | - | - |
| Rat Expression Set 230 | 1383034_at | - | - |
| Rat Expression Set 230 | 1383034_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864441 | - | - |
| Rat Genome U34 Set | rc_AA946264_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22303316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361370 | - | - |

Rybp
Genomic Map